Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-5590-5p |
---|---|
miRBase Accession Number: | MI0019150 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.2: 134857820 - 134857873 [+] on Build GRCh38 |
Mature miRNA Sequence: | UUGCCAUACAUAGACUUUAUU |
Species: | Human |
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UUGCCAUACAUAGACUUUAUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5590-5p | MIMAT0022299 |
Stem-loop ID | hsa-mir-5590 |
Stem-loop Accession # | MI0019150 |
Stem-loop Sequence | UUGCCAUACAUAGACUUUAUUGUGUUGAUCAACAAUAAAGUUCAUGUAUGGCAA |
Chromosome Location | Chr. 2 - 134857820 - 134857873 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022299
miRBase ID: hsa-miR-5590-5p Mature miRNA Sequence: UUGCCAUACAUAGACUUUAUU Chromosome Location: Chr. 2 - 134857820 - 134857873 [+] on Build GRCh38 |