Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-20a-3p |
---|---|
miRBase Accession Number: | MI0000076 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGCAUUAUGAGCACUUAAAG |
Species: | Human, Chrysemys picta |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCAUUAUGAGCACUUAAAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-20a-3p | MIMAT0004493 |
Chrysemys picta | cpi-miR-20a-3p | MIMAT0037648 |