Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › MH27694
            Assay Name: hsa-miR-6883-3p
            miRBase Accession Number: MI0022730
            miRBase Version: v22.1
            Chromosome Location: Chr.17: 8144994 - 8145071 [-] on Build GRCh38
            Mature miRNA Sequence: UUCCCUAUCUCACUCUCCUCAG
            Species: Human
            Product Type: mirVana® miRNA inhibitor
            Assay ID MH27694

            Purification
            Size
            Availability Made To Order
            Catalog # 4464084
            Price
            Your Price
            Online offer:
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Displaying...
            Build 38.2 Human Chr.17: MH27694×
            Created with Raphaël 2.1.2466405_mat480516_mir467209_mat480517_mirHs04348071_prihsa-mir-6883PER1MIR68838,141,4008,141,5008,141,6008,141,7008,141,8008,141,9008,142,0008,142,1008,142,2008,142,3008,142,4008,142,5008,142,6008,142,7008,142,8008,142,9008,143,0008,143,1008,143,2008,143,3008,143,4008,143,5008,143,6008,143,7008,143,8008,143,9008,144,0008,144,1008,144,2008,144,3008,144,4008,144,5008,144,6008,144,7008,144,8008,144,9008,145,0008,145,1008,145,2008,145,3008,145,4008,145,5008,145,6008,145,7008,145,8008,145,9008,146,0008,146,1008,146,2008,146,3008,146,4008,146,5008,146,6008,146,7008,146,8008,146,9008,147,0008,147,1008,147,2008,147,3008,147,4008,147,5008,147,6008,147,7008,147,8008,147,9008,148,0008,148,1008,148,2008,148,3008,148,4008,148,5008,148,600
            Back To Top

            Assay Details

            Mature miRNA Sequence:

            UUCCCUAUCUCACUCUCCUCAG

            miRBase Details:



            Species miRBase ID miRBase Accession Number
            Human hsa-miR-6883-3p MIMAT0027667

            Stem-loop Details



            Human

            Stem-loop ID hsa-mir-6883
            Stem-loop Accession # MI0022730
            Stem-loop Sequence
            CAGACAGGGAGGGUGUGGUAUGGAUGUGUUGACCCCUGAAGUGGUUCUGAUGACCUUUCCCUAUCUCACUCUCCUCAG
            Chromosome Location Chr. 17 - 8144994 - 8145071 [-] on Build GRCh38
            Mature MicroRNA miRBase Accession #: MIMAT0027667
            miRBase ID: hsa-miR-6883-3p
            Mature miRNA Sequence: UUCCCUAUCUCACUCUCCUCAG
            Chromosome Location: Chr. 17 - 8144994 - 8145071 [-] on Build GRCh38


            Back To Top

            More Information




            Related Products

            TaqMan™ Pri-miRNA Assay : Hs04348071_pri
            Ambion® Anti-miR™ miRNA Inhibitor : AM27694
            Ambion® Pre-miR™ miRNA Precursor : PM27694
            TaqMan™ MicroRNA Assay : 466405_mat
            mirVana® miRNA mimic : MC27694
            TaqMan™ Advanced miRNA Assay : 480516_mir

            Back To Top

            Related Products

            • Anti-GAPDH, Mouse Monoclonal 6C5
            • Anti-β-Actin, Mouse Monoclonal AC-15
            • TaqMan® Gene Expression Cells-to-CT™
            • Neon™ Transfection System 100 µL Kit
            • Lipofectamine™ RNAiMAX Transfection Reagent
            • Invivofectamine 2.0
            • Neon™ Transfection System
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-rms4r:80/100.66.77.126:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0