Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-18a-3p |
---|---|
miRBase Accession Number: | MI0000072 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGCCCUAAGUGCUCCUUCUGG |
Species: | Human, Cricetulus griseus, Goat |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCCCUAAGUGCUCCUUCUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-18a-3p | MIMAT0002891 |
Cricetulus griseus | cgr-miR-18a-3p | MIMAT0023818 |
Goat | chi-miR-18a-3p | MIMAT0036016 |