Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | mmu-miR-18a-3p |
---|---|
miRBase Accession Number: | MI0000567 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGCCCUAAGUGCUCCUUCUG |
Species: | Mouse, Alligator mississippiensis, Anolis carolinensis, Chicken, Chrysemys picta, Ophiophagus hannah, Python bivittatus |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCCCUAAGUGCUCCUUCUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-18a-3p | MIMAT0004626 |
Alligator mississippiensis | ami-miR-18-3p | MIMAT0038050 |
Anolis carolinensis | aca-miR-18a-3p | MIMAT0021818 |
Chicken | gga-miR-18a-3p | MIMAT0026499 |
Chrysemys picta | cpi-miR-18a-3p | MIMAT0037639 |
Ophiophagus hannah | oha-miR-18a-3p | MIMAT0036769 |
View More Rows | ||
Python bivittatus | pbv-miR-18a-3p | MIMAT0038841 |