Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | mmu-miR-7689-5p |
---|---|
Stem-loop Accession Number: | MI0025042 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.5: 36301570 - 36301628 [+] on Build GRCm38 |
Mature miRNA Sequence: | CCGAGGCAGCCUGGCUUAG |
Species: | Mouse |
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CCGAGGCAGCCUGGCUUAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-7689-5p | MIMAT0029908 |
Stem-loop ID | mmu-mir-7689 |
Stem-loop Accession # | MI0025042 |
Stem-loop Sequence | CCGAGGCAGCCUGGCUUAGUGGAAAAGGAACAGGCUUUAGAGCCAGACUGCCUGGGUUU |
Chromosome Location | Chr. 5 - 36301570 - 36301628 [+] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0029908
miRBase ID: mmu-miR-7689-5p Mature miRNA Sequence: CCGAGGCAGCCUGGCUUAG Chromosome Location: Chr. 5 - 36301570 - 36301628 [+] on Build GRCm38 |