Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
綜合式 CRO CDMO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
ITGA10
Entrez Gene ID
8515
Gene Name:
integrin subunit alpha 10
Gene Aliases
PRO827
Gene Chromosome Location
Chr.1: on Build GRCh38
UniGene
Hs.158237
Species:
Human
Primer Chromosome Location
Chr.1: 145893354-145892848 on Build GRCh38
View in UCSC browser
Amplicon Length
507
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00399893_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
ITGA10
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001303040.1
NP_001289969.1
-
-
-
NM_001303041.1
NP_001289970.1
-
-
-
NM_003637.4
NP_003628.2
5807607
c.3439-1G>T
p.?
6486678
c.3388G>T
p.V1130F
1294935
c.3349C>T
p.R1117W
24644
c.3415C>G
p.L1139V
4440828
c.3337G>C
p.V1113L
24643
c.3426C>T
p.F1142F
XM_005277436.3
XP_005277493.1
-
-
-
XM_011510083.2
XP_011508385.1
-
-
-
XM_011510084.1
XP_011508386.1
-
-
-
XM_017002622.1
XP_016858111.1
-
-
-
XM_017002623.1
XP_016858112.1
-
-
-
XM_017002624.1
XP_016858113.1
-
-
-
XM_017002625.1
XP_016858114.1
-
-
-
XM_017002626.1
XP_016858115.1
-
-
-
XM_017002628.1
XP_016858117.1
-
-
-
GenBank
AF074015.1
AAC31952.1
-
-
-
AF112345.1
AAF21944.1
-
-
-
AK304155.1
-
-
-
AK304183.1
-
-
-
AK314255.1
-
-
-
BC140831.1
-
-
-
BC144637.1
-
-
-
ENST00000369304.0
-
-
-
Phenotype
MIM: 604042
Literature Links:
ITGA10
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
integrin
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
integrin-mediated signaling pathway
extracellular matrix organization
collagen binding
metal ion binding
collagen binding involved in cell-matrix adhesion
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0