Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FGFR1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001174063.1 | NP_001167534.1 | - | - | - |
NM_001174064.1 | NP_001167535.1 | 6044172 | c.2156+8C>T | p.? | |
6928366 | c.2119C>T | p.H707Y | |||
5542923 | c.2152G>A | p.E718K | |||
6958491 | c.2154G>C | p.E718D | |||
6933849 | c.2122C>A | p.R708S | |||
6912534 | c.2101A>G | p.K701E | |||
5976839 | c.2136C>T | p.P712P | |||
4993070 | c.2102A>G | p.K701R | |||
6951649 | c.2094A>C | p.E698D | |||
6047851 | c.2123G>A | p.R708H | |||
6923959 | c.2092G>A | p.E698K | |||
1133829 | c.2109G>A | p.L703L | |||
NM_001174065.1 | NP_001167536.1 | - | - | - | |
NM_001174066.1 | NP_001167537.1 | - | - | - | |
NM_001174067.1 | NP_001167538.1 | - | - | - | |
NM_015850.3 | NP_056934.2 | - | - | - | |
NM_023105.2 | NP_075593.1 | - | - | - | |
NM_023106.2 | NP_075594.1 | - | - | - | |
NM_023110.2 | NP_075598.2 | 6044171 | c.2186+8C>T | p.? | |
422132 | c.2139G>A | p.L713L | |||
6047853 | c.2153G>A | p.R718H | |||
6912535 | c.2131A>G | p.K711E | |||
6923958 | c.2122G>A | p.E708K | |||
6191515 | c.2156T>C | p.M719T | |||
5542922 | c.2182G>A | p.E728K | |||
6958490 | c.2184G>C | p.E728D | |||
5976841 | c.2166C>T | p.P722P | |||
6923960 | c.2122G>A | p.E708K | |||
6933848 | c.2152C>A | p.R718S | |||
6958492 | c.2184G>C | p.E728D | |||
6047850 | c.2153G>A | p.R718H | |||
6933850 | c.2152C>A | p.R718S | |||
6044174 | c.2186+8C>T | p.? | |||
7412626 | c.2143G>T | p.E715* | |||
5976838 | c.2166C>T | p.P722P | |||
4993069 | c.2132A>G | p.K711R | |||
5542924 | c.2182G>A | p.E728K | |||
6951648 | c.2124A>C | p.E708D | |||
422133 | c.2139G>A | p.L713L | |||
422134 | c.2133+6G>A | p.? | |||
6951650 | c.2124A>C | p.E708D | |||
6928365 | c.2149C>T | p.H717Y | |||
6912533 | c.2131A>G | p.K711E | |||
6928367 | c.2149C>T | p.H717Y | |||
4993071 | c.2132A>G | p.K711R | |||
XM_006716303.2 | XP_006716366.1 | - | - | - | |
XM_006716304.1 | XP_006716367.1 | - | - | - | |
XM_006716306.2 | XP_006716369.1 | - | - | - | |
XM_006716307.1 | XP_006716370.1 | - | - | - | |
XM_006716309.3 | XP_006716372.1 | - | - | - | |
XM_006716310.2 | XP_006716373.1 | - | - | - | |
XM_006716311.1 | XP_006716374.1 | - | - | - | |
XM_006716312.1 | XP_006716375.1 | - | - | - | |
XM_006716313.2 | XP_006716376.1 | - | - | - | |
XM_006716314.1 | XP_006716377.1 | - | - | - | |
XM_011544443.2 | XP_011542745.1 | - | - | - | |
XM_011544444.1 | XP_011542746.1 | - | - | - | |
XM_011544445.2 | XP_011542747.1 | - | - | - | |
XM_011544446.2 | XP_011542748.1 | - | - | - | |
XM_011544447.2 | XP_011542749.1 | - | - | - | |
XM_011544448.1 | XP_011542750.1 | - | - | - | |
XM_011544449.1 | XP_011542751.1 | - | - | - | |
XM_011544450.2 | XP_011542752.1 | - | - | - | |
XM_011544451.1 | XP_011542753.1 | - | - | - | |
XM_017013219.1 | XP_016868708.1 | - | - | - | |
XM_017013220.1 | XP_016868709.1 | - | - | - | |
XM_017013221.1 | XP_016868710.1 | - | - | - | |
XM_017013222.1 | XP_016868711.1 | - | - | - | |
XM_017013223.1 | XP_016868712.1 | - | - | - | |
XM_017013224.1 | XP_016868713.1 | - | - | - | |
XM_017013225.1 | XP_016868714.1 | - | - | - | |
XM_017013226.1 | XP_016868715.1 | - | - | - | |
XM_017013227.1 | XP_016868716.1 | - | - | - | |
XM_017013228.1 | XP_016868717.1 | - | - | - | |
XM_017013229.1 | XP_016868718.1 | - | - | - | |
XM_017013230.1 | XP_016868719.1 | - | - | - | |
GenBank | AB208919.1 | BAD92156.1 | - | - | - |
AK222718.1 | BAD96438.1 | - | - | - | |
AK291754.1 | - | - | - | ||
AK292470.1 | - | - | - | ||
AK309947.1 | - | - | - | ||
BC015035.1 | AAH15035.1 | - | - | - | |
BC018128.1 | AAH18128.1 | - | - | - | |
BC091494.1 | AAH91494.1 | - | - | - | |
BX649167.1 | - | - | - | ||
ENST00000447712.0 | - | - | - | ||
FJ809917.1 | - | - | - | ||
KU178066.1 | - | - | - | ||
KU178067.1 | - | - | - | ||
KU178068.1 | - | - | - | ||
KU178069.1 | - | - | - | ||
KU178070.1 | - | - | - | ||
M34185.1 | AAA35836.1 | - | - | - | |
M34186.1 | AAA35837.1 | - | - | - | |
M34641.1 | AAA35835.1 | - | - | - | |
M37722.1 | AAA75007.1 | - | - | - | |
M60485.1 | AAA35840.1 | - | - | - | |
M63887.1 | AAA35958.1 | - | - | - | |
M63888.1 | AAA35959.1 | - | - | - | |
X51803.1 | CAA36101.1 | - | - | - | |
X52833.1 | CAA37015.1 | - | - | - | |
X57119.1 | CAA40401.1 | - | - | - | |
X57120.1 | CAA40402.1 | - | - | - | |
X57121.1 | CAA40403.1 | - | - | - | |
X57122.1 | CAA40404.1 | - | - | - | |
X66945.1 | CAA47375.1 | - | - | - | |
Y00665.1 | CAA68679.1 | - | - | - |
Literature Links: |
FGFR1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |