Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GRM8 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000845.2 | NP_000836.2 | 141888 | c.1946C>T | p.S649F |
351509 | c.1957G>C | p.V653L | |||
2768119 | c.2024G>A | p.R675Q | |||
6515557 | c.2088G>T | p.L696L | |||
2152664 | c.2030T>C | p.F677S | |||
3366763 | c.2073T>C | p.S691S | |||
6108779 | c.2011A>T | p.N671Y | |||
3632826 | c.2094C>A | p.I698I | |||
1192300 | c.2015G>A | p.R672H | |||
7089642 | c.2037G>T | p.Q679H | |||
744715 | c.1994C>A | p.A665D | |||
4391924 | c.2044A>G | p.K682E | |||
166590 | c.2057C>T | p.A686V | |||
6176338 | c.1955G>T | p.R652L | |||
94423 | c.2072G>C | p.S691T | |||
6515559 | c.2058G>T | p.A686A | |||
4475022 | c.1959C>T | p.V653V | |||
3632828 | c.2081C>A | p.S694Y | |||
7268982 | c.2084A>G | p.Q695R | |||
6108781 | c.2096C>A | p.T699N | |||
77816 | c.2028A>G | p.I676M | |||
2768121 | c.1972G>C | p.G658R | |||
6193583 | c.2010A>C | p.T670T | |||
203268 | c.1952G>A | p.R651Q | |||
744716 | c.2043G>A | p.K681K | |||
333059 | c.2038G>T | p.G680W | |||
3949874 | c.2014C>A | p.R672S | |||
3922631 | c.2020C>T | p.H674Y | |||
5532073 | c.2094C>T | p.I698I | |||
6176340 | c.2050G>A | p.V684I | |||
6515561 | c.1974C>T | p.G658G | |||
7370418 | c.2074C>A | p.P692T | |||
94424 | c.2007A>T | p.K669N | |||
4475809 | c.2019C>T | p.I673I | |||
744717 | c.2058G>C | p.A686A | |||
5630939 | c.1964T>C | p.L655P | |||
3632832 | c.2004C>T | p.T668T | |||
3877802 | c.1934C>A | p.T645K | |||
4445084 | c.2023C>T | p.R675* | |||
6210890 | c.1986C>G | p.S662R | |||
3784973 | c.1955G>A | p.R652Q | |||
1662083 | c.2065T>A | p.F689I | |||
6176342 | c.2071_2072AG>TT | p.S691F | |||
2768118 | c.2040G>A | p.G680G | |||
6548704 | c.1995C>A | p.A665A | |||
6889096 | c.1947C>T | p.S649S | |||
5976208 | c.1975A>G | p.M659V | |||
128114 | c.2091G>A | p.V697V | |||
4533955 | c.2032G>A | p.E678K | |||
6676227 | c.2001G>A | p.L667L | |||
4689004 | c.2018T>G | p.I673S | |||
6478935 | c.1959C>A | p.V653V | |||
7245688 | c.1954C>T | p.R652W | |||
4475877 | c.2022C>T | p.H674H | |||
6108777 | c.1986C>T | p.S662S | |||
5738137 | c.1978T>C | p.C660R | |||
6676219 | c.1973G>T | p.G658V | |||
NM_001127323.1 | NP_001120795.1 | 6515560 | c.2058G>T | p.A686A | |
7370419 | c.2074C>A | p.P692T | |||
4475878 | c.2022C>T | p.H674H | |||
6478936 | c.1959C>A | p.V653V | |||
6108778 | c.1986C>T | p.S662S | |||
3698018 | c.1952G>A | p.R651Q | |||
7268983 | c.2084A>G | p.Q695R | |||
3632831 | c.2024G>A | p.R675Q | |||
4445085 | c.2023C>T | p.R675* | |||
5532074 | c.2094C>T | p.I698I | |||
6889097 | c.1947C>T | p.S649S | |||
4862153 | c.2058G>C | p.A686A | |||
6176341 | c.2050G>A | p.V684I | |||
3949875 | c.2014C>A | p.R672S | |||
6210891 | c.1986C>G | p.S662R | |||
6515562 | c.1974C>T | p.G658G | |||
4861514 | c.1957G>C | p.V653L | |||
6108780 | c.2011A>T | p.N671Y | |||
3877801 | c.2015G>A | p.R672H | |||
6676228 | c.2001G>A | p.L667L | |||
4689005 | c.2018T>G | p.I673S | |||
3632833 | c.2004C>T | p.T668T | |||
7089643 | c.2037G>T | p.Q679H | |||
6176343 | c.2071_2072AG>TT | p.S691F | |||
3922632 | c.2020C>T | p.H674Y | |||
4862172 | c.1994C>A | p.A665D | |||
3366764 | c.2073T>C | p.S691S | |||
3632827 | c.2094C>A | p.I698I | |||
4533956 | c.2032G>A | p.E678K | |||
6108782 | c.2096C>A | p.T699N | |||
7245689 | c.1954C>T | p.R652W | |||
4475023 | c.1959C>T | p.V653V | |||
3877803 | c.1934C>A | p.T645K | |||
3632834 | c.1946C>T | p.S649F | |||
6676220 | c.1973G>T | p.G658V | |||
4359488 | c.2057C>T | p.A686V | |||
5976209 | c.1975A>G | p.M659V | |||
3784974 | c.1955G>A | p.R652Q | |||
3632829 | c.2081C>A | p.S694Y | |||
5738138 | c.1978T>C | p.C660R | |||
6548705 | c.1995C>A | p.A665A | |||
4861213 | c.2043G>A | p.K681K | |||
6515558 | c.2088G>T | p.L696L | |||
5630940 | c.1964T>C | p.L655P | |||
4475810 | c.2019C>T | p.I673I | |||
3411544 | c.2030T>C | p.F677S | |||
4359491 | c.1972G>C | p.G658R | |||
4391925 | c.2044A>G | p.K682E | |||
6176339 | c.1955G>T | p.R652L | |||
3632830 | c.2040G>A | p.G680G | |||
6193584 | c.2010A>C | p.T670T | |||
NR_028041.1 | - | - | - | ||
XM_006715938.3 | XP_006716001.1 | - | - | - | |
XM_011516091.1 | XP_011514393.1 | - | - | - | |
XM_011516092.2 | XP_011514394.1 | - | - | - | |
XM_011516094.1 | XP_011514396.1 | - | - | - | |
XM_011516095.2 | XP_011514397.1 | - | - | - | |
XM_011516101.2 | XP_011514403.1 | - | - | - | |
XM_011516102.2 | XP_011514404.1 | - | - | - | |
XM_011516103.2 | XP_011514405.1 | - | - | - | |
XM_011516108.2 | XP_011514410.1 | - | - | - | |
XM_017012074.1 | XP_016867563.1 | - | - | - | |
XM_017012075.1 | XP_016867564.1 | - | - | - | |
XM_017012076.1 | XP_016867565.1 | - | - | - | |
XM_017012077.1 | XP_016867566.1 | - | - | - | |
XM_017012078.1 | XP_016867567.1 | - | - | - | |
XM_017012079.1 | XP_016867568.1 | - | - | - | |
XM_017012080.1 | XP_016867569.1 | - | - | - | |
GenBank | AJ236921.1 | CAB36968.1 | - | - | - |
AJ236922.1 | CAB36969.1 | - | - | - | |
AK123053.1 | - | - | - | ||
AK290197.1 | - | - | - | ||
AK315203.1 | - | - | - | ||
AY608335.1 | AAT37959.1 | - | - | - | |
BC093725.1 | AAH93725.1 | - | - | - | |
BC101675.1 | AAI01676.1 | - | - | - | |
BC143344.1 | - | - | - | ||
ENST00000339582.0 | - | - | - | ||
EU432125.1 | - | - | - | ||
U92459.1 | AAB51764.1 | - | - | - | |
U95025.1 | AAB72040.1 | - | - | - |
Literature Links: |
GRM8 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |