Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            Gene Symbol:
            GEMIN2
            Entrez Gene ID
            8487
            Gene Name:
            gem nuclear organelle associated protein 2
            Gene Aliases
            SIP1, SIP1-delta
            Gene Chromosome Location
            Chr.14: on Build GRCh38
            UniGene
            Hs.652307
            Species:
            Human
            Primer Chromosome Location
            Chr.14: 39118433-39118694 on Build GRCh38 View in UCSC browser
            Amplicon Length
            262
            Primer Sequences
            Log in to view
            Ampliseq ID's
            Primer ID Hs00562604_CE
            Primer 5'-tail Option
            Purification
            Primer Pair /FWD/REV
            Synthesis Scale 25 nmol
            Availability Made To Order
            Catalog #'s A15629,A15630
            Price (USD) USD 16.46,USD 16.46
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data...
            ×
            Back To Top

            Assay Details

            Gene Information


            GEMIN2
            Accession Protein COSMIC ID Mutation AA
            RefSeq NM_001009182.1 NP_001009182.1 - - -
            NM_001009183.1 NP_001009183.1 - - -
            NM_003616.2 NP_003607.1 6670412 c.405+2T>C p.?
            XM_017021709.1 XP_016877198.1 - - -
            GenBank AB037701.1 BAB03508.1 - - -
            AB037702.1 BAB03509.1 - - -
            AB037703.1 BAB03510.1 - - -
            AF027150.1 AAB82297.1 - - -
            AK313947.1 - - -
            BC028095.1 - - -
            BC104968.1 AAI04969.1 - - -
            CN484054.1 - - -


            Phenotype

            MIM: 602595

            Literature Links:

            GEMIN2 PubMed Links

            SNP ID

            rs775565207 rs556211192 rs765241854 rs970278916 rs762623063 rs966982616 rs751907017 rs762071534 rs376851598 rs189184432 rs753212010 rs372555968 rs781704765 rs934645610 rs569293506 rs527454695 rs777299234 rs369766094 rs139047393 rs990411386 rs192271975 rs960252811 rs750474827

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118517-39118517 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118628-39118628 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            G (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            G (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            G (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            G (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            G (0.00)
            (1.00)
            AMR
            G (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118523-39118524 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118598-39118598 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118536-39118536 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118493-39118493 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118579-39118579 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118645-39118645 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118519-39118519 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118603-39118603 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            A (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            A (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            A (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            A (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            A (0.00)
            (1.00)
            AMR
            A (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118616-39118616 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118502-39118502 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118612-39118612 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118597-39118597 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118650-39118650 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            C (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            C (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            C (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            C (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            C (0.00)
            (1.00)
            AMR
            C (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118465-39118465 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            G (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            G (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            G (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            G (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            G (0.00)
            (1.00)
            AMR
            G (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118633-39118633 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118569-39118569 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            G (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            G (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            G (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            G (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            G (0.00)
            (1.00)
            AMR
            G (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118594-39118594 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            T (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            T (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            T (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            T (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            T (0.00)
            (1.00)
            AMR
            T (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118578-39118578 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118668-39118668 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            C (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            C (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            C (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            C (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            C (0.00)
            (1.00)
            AMR
            C (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118492-39118492 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.14: 39118553-39118553 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available


            Back To Top

            More Information


            Set Membership:

            Between 200-400 bp

            Panther Classification:

            Molecular Function -

            RNA splicing factor

            Gene Ontology Categories:

            Function(s) Process(es)

            spliceosomal complex assembly
            RNA splicing, via transesterification reactions
            spliceosomal snRNP assembly
            mRNA processing
            RNA splicing
            nuclear import
            protein binding

            Format:

            Each primer will be delivered in a single tube, dried-down

            Quick Reference Guide

            Primer Designer™ Tool Quick Reference




            Back To Top

            Related Products

            • POP-6™ Polymer
            • POP-7™ Polymer
            • BigDye® Terminator v1.1 Cycle Sequencing Kit
            • BigDye® Terminator v3.1 Cycle Sequencing Kit
            • BigDye® Direct Cycle Sequencing Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-sbk6v:80/100.66.72.122:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline