Save up to 31% on select cell culture essentials

Bundle and save

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            Gene Symbol:
            CHRM1
            Entrez Gene ID
            1128
            Gene Name:
            cholinergic receptor muscarinic 1
            Gene Aliases
            HM1, M1, M1R
            Gene Chromosome Location
            Chr.11: on Build GRCh38
            UniGene
            Hs.632119
            Species:
            Human
            Primer Chromosome Location
            Chr.11: 62910442-62910679 on Build GRCh38 View in UCSC browser
            Amplicon Length
            238
            Primer Sequences
            Log in to view
            Ampliseq ID's
            Primer ID Hs00565941_CE
            Primer 5'-tail Option
            Purification
            Primer Pair /FWD/REV
            Synthesis Scale 25 nmol
            Availability Made To Order
            Catalog #'s A15629,A15630
            Price
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data.
            ×
            Back To Top

            Assay Details

            Gene Information


            CHRM1
            Accession Protein COSMIC ID Mutation AA
            RefSeq NM_000738.2 NP_000729.2 2162049 c.617C>T p.T206M
            4412322 c.511C>T p.R171W
            7272373 c.637C>T p.R213W
            4756397 c.659G>A p.R220Q
            3451126 c.549C>T p.L183L
            6321738 c.567C>A p.T189T
            3451127 c.540C>T p.I180I
            6007741 c.617C>A p.T206K
            6230354 c.422G>A p.R141Q
            6880579 c.512G>A p.R171Q
            5761877 c.657A>G p.A219A
            291976 c.588C>G p.A196A
            6250497 c.493C>A p.Q165K
            1355487 c.446C>T p.A149V
            6269874 c.515C>A p.T172K
            6133986 c.471G>T p.W157C
            6133987 c.558C>A p.P186P
            5739294 c.439G>T p.G147C
            XM_011544742.2 XP_011543044.1 - - -
            GenBank AF385587.1 AAK68112.1 - - -
            AF498915.1 AAM18938.1 - - -
            AK312374.1 - - -
            BC007740.2 AAH07740.1 - - -
            BC022984.1 AAH22984.1 - - -
            BT007166.1 AAP35830.1 - - -
            ENST00000306960.0 - - -


            Phenotype

            MIM: 118510

            Literature Links:

            CHRM1 PubMed Links

            SNP ID

            rs1049259397 rs774456975 rs146879247 rs1045782221 rs765546937 rs755589700 rs750443392 rs753270791 rs374933200 rs768496909 rs559478266 rs200340019 rs139087623 rs755342464 rs371629017 rs761028069 rs555027009 rs754003908 rs201681850 rs766578516 rs539642013

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910546-62910546 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910470-62910470 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910463-62910463 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910472-62910472 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910573-62910573 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910648-62910648 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910484-62910484 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910595-62910595 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910477-62910477 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910464-62910464 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910594-62910594 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910606-62910606 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            T (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            T (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            T (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            T (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            T (0.00)
            (1.00)
            AMR
            T (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910483-62910483 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            T (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            T (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            T (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            T (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            T (0.00)
            (1.00)
            AMR
            T (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910549-62910549 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910471-62910471 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            A (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            A (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            A (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            A (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            A (0.00)
            (1.00)
            AMR
            A (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910494-62910494 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910633-62910633 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910539-62910539 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910627-62910627 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            C (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            C (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            C (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            C (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            C (0.00)
            (1.00)
            AMR
            C (0.00)
            (1.00)

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910537-62910537 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
            EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
            SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
            EUR - Not Available
            AMR - Not Available

            Submission count

            NA

            Allele Nomenclature



            Chromosome Location

            Chr.11: 62910660-62910660 on Build GRCh38

            Allele Frequency



            1000Genome Applied Biosystems® HapMap
            Global
            A (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            A (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            A (0.00)
            (1.00)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            A (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            A (0.00)
            (1.00)
            AMR
            A (0.00)
            (1.00)


            Back To Top

            More Information


            Set Membership:

            Between 200-400 bp

            Panther Classification:

            Molecular Function -

            G-protein coupled receptor

            Gene Ontology Categories:

            Function(s) Process(es)

            regulation of vascular smooth muscle contraction
            cellular protein modification process
            signal transduction
            G-protein coupled receptor signaling pathway
            adenylate cyclase-inhibiting G-protein coupled acetylcholine receptor signaling pathway
            protein kinase C-activating G-protein coupled receptor signaling pathway
            phospholipase C-activating G-protein coupled acetylcholine receptor signaling pathway
            G-protein coupled acetylcholine receptor signaling pathway
            synaptic transmission, cholinergic
            neuromuscular synaptic transmission
            nervous system development
            cell proliferation
            positive regulation of cell proliferation
            regulation of locomotion
            positive regulation of ion transport
            saliva secretion
            cognition
            positive regulation of intracellular protein transport
            phosphatidylinositol phospholipase C activity
            drug binding
            G-protein coupled acetylcholine receptor activity

            Format:

            Each primer will be delivered in a single tube, dried-down

            Quick Reference Guide

            Primer Designer™ Tool Quick Reference




            Back To Top

            Related Products

            • POP-6™ Polymer
            • POP-7™ Polymer
            • BigDye® Terminator v1.1 Cycle Sequencing Kit
            • BigDye® Terminator v3.1 Cycle Sequencing Kit
            • BigDye® Direct Cycle Sequencing Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-srd2g:80/100.66.79.97:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline