Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
KCNMA1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001014797.2 | NP_001014797.1 | - | - | - |
NM_001161352.1 | NP_001154824.1 | - | - | - | |
NM_001161353.1 | NP_001154825.1 | - | - | - | |
NM_001271518.1 | NP_001258447.1 | - | - | - | |
NM_001271519.1 | NP_001258448.1 | - | - | - | |
NM_001322829.1 | NP_001309758.1 | - | - | - | |
NM_001322830.1 | NP_001309759.1 | - | - | - | |
NM_001322832.1 | NP_001309761.1 | - | - | - | |
NM_001322835.1 | NP_001309764.1 | - | - | - | |
NM_001322836.1 | NP_001309765.1 | - | - | - | |
NM_001322837.1 | NP_001309766.1 | - | - | - | |
NM_001322838.1 | NP_001309767.1 | 7303577 | c.2408C>T | p.P803L | |
4774788 | c.2345T>G | p.V782G | |||
5508625 | c.2358C>G | p.C786W | |||
6883523 | c.2321G>A | p.W774* | |||
6518027 | c.2328G>T | p.P776P | |||
6883524 | c.2424G>A | p.K808K | |||
5703711 | c.2415C>A | p.S805R | |||
1349213 | c.2354C>T | p.T785I | |||
4536708 | c.2335G>A | p.G779R | |||
7298801 | c.2318G>A | p.G773D | |||
1965782 | c.2411C>T | p.A804V | |||
NM_002247.3 | NP_002238.2 | - | - | - | |
XM_005269776.3 | XP_005269833.1 | - | - | - | |
XM_005269778.1 | XP_005269835.1 | - | - | - | |
XM_005269781.2 | XP_005269838.1 | - | - | - | |
XM_005269787.3 | XP_005269844.1 | - | - | - | |
XM_005269789.2 | XP_005269846.1 | - | - | - | |
XM_005269792.2 | XP_005269849.1 | - | - | - | |
XM_005269796.2 | XP_005269853.1 | - | - | - | |
XM_006717826.2 | XP_006717889.1 | - | - | - | |
XM_011539773.1 | XP_011538075.1 | - | - | - | |
XM_011539774.1 | XP_011538076.1 | - | - | - | |
XM_011539775.1 | XP_011538077.1 | - | - | - | |
XM_011539776.1 | XP_011538078.1 | - | - | - | |
XM_011539777.1 | XP_011538079.1 | - | - | - | |
XM_011539778.2 | XP_011538080.1 | - | - | - | |
XM_011539779.1 | XP_011538081.1 | - | - | - | |
XM_011539780.1 | XP_011538082.1 | - | - | - | |
XM_011539781.2 | XP_011538083.1 | - | - | - | |
XM_011539782.2 | XP_011538084.1 | - | - | - | |
XM_011539783.1 | XP_011538085.1 | - | - | - | |
XM_011539785.1 | XP_011538087.1 | - | - | - | |
XM_017016207.1 | XP_016871696.1 | - | - | - | |
XM_017016208.1 | XP_016871697.1 | - | - | - | |
XM_017016209.1 | XP_016871698.1 | - | - | - | |
XM_017016210.1 | XP_016871699.1 | - | - | - | |
XM_017016211.1 | XP_016871700.1 | - | - | - | |
XM_017016212.1 | XP_016871701.1 | - | - | - | |
XM_017016213.1 | XP_016871702.1 | - | - | - | |
XM_017016214.1 | XP_016871703.1 | - | - | - | |
XM_017016215.1 | XP_016871704.1 | - | - | - | |
XM_017016216.1 | XP_016871705.1 | - | - | - | |
XM_017016217.1 | XP_016871706.1 | - | - | - | |
XM_017016218.1 | XP_016871707.1 | - | - | - | |
XM_017016222.1 | XP_016871711.1 | - | - | - | |
XM_017016223.1 | XP_016871712.1 | - | - | - | |
GenBank | AB113382.1 | BAD06365.1 | - | - | - |
AB113575.1 | BAD06397.1 | - | - | - | |
AB209488.1 | BAD92725.1 | - | - | - | |
AB524033.1 | - | - | - | ||
AF025999.1 | AAB88802.1 | - | - | - | |
AF118141.1 | AAD31173.1 | - | - | - | |
AK310379.1 | - | - | - | ||
AY040849.1 | AAK91504.1 | - | - | - | |
BC137115.1 | - | - | - | ||
BC137137.1 | - | - | - | ||
BC144496.1 | - | - | - | ||
BX648925.1 | - | - | - | ||
ENST00000286627.0 | - | - | - | ||
U02632.1 | AAA50173.1 | - | - | - | |
U11058.2 | AAB65837.1 | - | - | - | |
U11717.1 | AAC50353.1 | - | - | - | |
U13913.1 | AAA85104.1 | - | - | - | |
U23767.1 | AAA92290.1 | - | - | - |
KCNMA1-AS1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NR_120655.1 | - | - | - | |
GenBank | BX092192.1 | - | - | - |
Literature Links: |
KCNMA1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |