Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ANGPTL8 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_018687.6 | NP_061157.3 | 5527623 | c.426G>A | p.L142L |
7358192 | c.439C>G | p.R147G | |||
4805428 | c.444A>C | p.E148D | |||
990846 | c.438C>T | p.Y146Y | |||
6614981 | c.449A>G | p.E150G | |||
4935936 | c.386G>T | p.S129I | |||
4987328 | c.364G>C | p.A122P | |||
GenBank | AF271350.2 | AAF76204.2 | - | - | - |
AY358356.1 | AAQ88722.1 | - | - | - | |
BC113823.1 | AAI13824.1 | - | - | - | |
BP316755.1 | - | - | - | ||
KF809856.1 | - | - | - | ||
KF809857.1 | - | - | - | ||
KF809858.1 | - | - | - |
DOCK6 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_020812.3 | NP_065863.2 | - | - | - |
XM_005260000.2 | XP_005260057.1 | - | - | - | |
XM_005260001.2 | XP_005260058.1 | - | - | - | |
XM_011528150.1 | XP_011526452.1 | - | - | - | |
XM_011528151.1 | XP_011526453.1 | - | - | - | |
XM_011528152.1 | XP_011526454.1 | - | - | - | |
GenBank | AB037816.2 | - | - | - | |
BC146786.1 | - | - | - |
Literature Links: |
ANGPTL8 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |