- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 1 | 634 | ||
NM_001318368.1 | NP_001305297.1 | 1 | 634 | |||
XM_011534864.1 | XP_011533166.1 | 1 | 613 | |||
XM_017020317.1 | XP_016875806.1 | 1 | 66 | |||
GenBank mRNA | AY702976.1 | - | 1 | 96 | ||
BC069634.1 | - | 1 | 119 | 7262421 | ||
BC094784.1 | - | 1 | 634 | |||
BC104492.1 | - | 1 | 122 | 40032854 | ||
BC105950.1 | - | 1 | 122 | 40032852 | ||
BC107601.1 | - | 1 | 121 | 40032849 | ||
X85753.1 | - | 1 | 122 |