Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178726.3 | NP_848841.2 | 1 | 843 | ||
GenBank mRNA | AK032529.1 | - | 1 | 901 | ||
AK035912.1 | - | 1 | 846 | |||
AK045724.1 | - | 1 | 884 | |||
AK131646.1 | - | 1 | 830 | |||
AK142789.1 | - | 1 | 861 | |||
AK147389.1 | - | 1 | 846 | |||
AK147876.1 | - | 1 | 880 | |||
AK165160.1 | - | 1 | 876 | |||
AK220523.1 | - | 1 | 848 | |||
AY184801.1 | - | 1 | 431 | |||
BC096031.1 | - | 1 | 843 | 6837066 |