Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289836.1 | NP_001276765.1 | 3 | 643 | ||
NM_001289837.1 | NP_001276766.1 | 3 | 617 | |||
NM_001289838.1 | NP_001276767.1 | 3 | 324 | |||
NM_001289839.1 | NP_001276768.1 | 3 | 243 | |||
NM_178734.4 | NP_848849.2 | 3 | 469 | |||
GenBank mRNA | AK030093.1 | - | 3 | 469 | ||
AK036453.1 | - | 3 | 282 | |||
AK050747.1 | - | 3 | 317 | |||
AK173109.1 | - | 2 | 2568 | |||
BC096555.1 | - | 3 | 241 | 30614720 | ||
DV657711.1 | - | 486 |