Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178778.3 | NP_848893.2 | 2, 3 | 194 | ||
GenBank mRNA | AK044772.1 | - | 2, 3 | 190 | ||
AK083280.1 | - | 2, 3 | 196 | |||
AK163712.1 | - | 2, 3 | 198 | |||
BC132485.1 | - | 2, 3 | 169 | 40130762 | ||
BY783760.1 | - | 2, 3 | 198 |