Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IFIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001547.4 | NP_001538.4 | 2 | 447 | ||
GenBank mRNA | AK300431.1 | - | 2 | 349 | ||
AK312831.1 | - | 2 | 396 | |||
BC005987.1 | - | 2 | 350 | |||
BC032839.2 | - | 2 | 350 | 4838844 | ||
DA631671.1 | - | 2 | 451 |