Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289836.1 | NP_001276765.1 | 3, 4 | 742 | ||
NM_001289837.1 | NP_001276766.1 | 3, 4 | 716 | |||
NM_178734.4 | NP_848849.2 | 3, 4 | 568 | |||
GenBank mRNA | AK030093.1 | - | 3, 4 | 568 | ||
AK050747.1 | - | 3, 4 | 416 | |||
AK173109.1 | - | 2, 3 | 2667 | |||
DV657711.1 | - | 585 |