Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egflam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289496.1 | NP_001276425.1 | 3, 4 | 590 | ||
NM_001289498.1 | NP_001276427.1 | 3, 4 | 590 | |||
NM_178748.6 | NP_848863.1 | 3, 4 | 590 | |||
GenBank mRNA | AK031136.1 | - | 3, 4 | 499 | ||
AK033332.1 | - | 3, 4 | 507 | |||
AK037223.1 | - | 3, 4 | 492 | |||
AK041546.1 | - | 3, 4 | 471 | |||
AK087659.1 | - | 3, 4 | 470 | |||
AK158909.1 | - | 3, 4 | 591 | |||
BC095994.1 | - | 3, 4 | 320 | 6467602 | ||
BC150710.1 | - | 3, 4 | 310 | 9087621 | ||
DQ223720.1 | - | 3, 4 | 294 |