Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egflam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289496.1 | NP_001276425.1 | 23 | 4400 | ||
NM_001289498.1 | NP_001276427.1 | 23 | 4395 | |||
NM_178748.6 | NP_848863.1 | 22 | 4376 | |||
GenBank mRNA | AK031136.1 | - | 22 | 4285 | ||
AK033332.1 | - | 22 | 4293 | |||
AK158909.1 | - | 23 | 4397 | |||
BC051455.1 | - | 8 | 1929 | |||
DQ223720.1 | - | 23 | 4104 |