Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178877.6 | NP_849208.4 | 13 | 1930 | ||
GenBank mRNA | AK029417.1 | - | 10 | 1481 | ||
AK039655.1 | - | 4 | 929 | |||
BC090977.1 | - | 13 | 1883 | 30503721 |