Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009169.2 | NP_033195.1 | 2 | 232 | ||
GenBank mRNA | AK135462.1 | - | 185 | |||
AK160432.1 | - | 2 | 225 | |||
AK161690.1 | - | 2 | 205 | |||
AK167914.1 | - | 2 | 182 | |||
BC023490.1 | - | 2 | 189 | 5251089 | ||
BC058117.1 | - | 2 | 198 | 6412144 | ||
U41626.1 | - | 2 | 174 | |||
X83589.1 | - | 2 | 208 |