Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177411.4 | NP_803130.1 | 2 | 316 | ||
GenBank mRNA | AB232594.1 | - | 1 | 177 | ||
AK081251.1 | - | 2 | 312 | |||
AK083034.1 | - | 2 | 305 | |||
AK144551.1 | - | 2 | 438 | |||
BC137574.1 | - | 1 | 228 | 8860593 | ||
BC137575.1 | - | 1 | 228 | 8860594 | ||
BY727964.1 | - | 2 | 317 | |||
M79311.1 | - | 1 | 42 | |||
X84239.1 | - | 1 | 185 |