Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011873.2 | NP_036003.2 | 2 | 151 | ||
GenBank mRNA | AF085348.1 | - | 2 | 116 | ||
AK002595.1 | - | 2 | 188 | |||
AK050159.1 | - | 2 | 165 | |||
AK088916.1 | - | 2 | 173 | |||
AK132139.1 | - | 2 | 165 | |||
AK137246.1 | - | 2 | 152 | |||
AK145398.1 | - | 2 | 185 | |||
AK146970.1 | - | 2 | 176 | |||
AK151730.1 | - | 1 | 867 | |||
AK152511.1 | - | 1 | 867 | |||
AK169769.1 | - | 1 | 451 | |||
BC014808.1 | - | 2 | 151 | 4506124 | ||
BC029351.1 | - | 1 | 82 | |||
BC083347.1 | - | 2 | 143 | 6829350 |