Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009609.2 | NP_033739.1 | 4 | 691 | ||
GenBank mRNA | AF195094.1 | - | 4 | 721 | ||
AK012251.1 | - | 4 | 703 | |||
AK076081.1 | - | 4 | 700 | |||
AK087983.1 | - | 4 | 700 | |||
AK149927.1 | - | 4 | 697 | |||
AK150209.1 | - | 4 | 697 | |||
AK152202.1 | - | 4 | 704 | |||
AK153446.1 | - | 4 | 702 | |||
AK157726.1 | - | 4 | 700 | |||
AK159867.1 | - | 4 | 703 | |||
AK162157.1 | - | 4 | 697 | |||
AK200871.1 | - | 1 | 213 | |||
AK208587.1 | - | 1 | 79 | |||
AK208588.1 | - | 1 | 79 | |||
AK209715.1 | - | 1 | 79 | |||
BC003337.1 | - | 4 | 686 | 3257312 | ||
BC018571.1 | - | 4 | 686 | |||
BC021796.1 | - | 4 | 688 | 4218945 | ||
BC023248.1 | - | 4 | 691 | 3499390 | ||
BC099371.1 | - | 4 | 683 | 30841085 | ||
M21495.1 | - | 4 | 664 | |||
X13055.1 | - | 3 | 606 |