Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018853.3 | NP_061341.1 | 3 | 396 | ||
GenBank mRNA | AK007832.1 | - | 3 | 427 | ||
AK010656.1 | - | 3 | 428 | |||
AK088070.1 | - | 3 | 426 | |||
AK169347.1 | - | 3 | 426 | |||
AK203941.1 | - | 1 | 112 | |||
BC013252.1 | - | 3 | 361 | |||
BC058685.1 | - | 3 | 438 | 6410847 | ||
BC091747.1 | - | 3 | 401 | 2655117 | ||
BC092088.1 | - | 3 | 400 | 5102711 | ||
BC092536.1 | - | 396 | 6774435 | |||
U29402.1 | - | 3 | 424 |