Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177411.4 | NP_803130.1 | 3 | 426 | ||
GenBank mRNA | AB232594.1 | - | 2 | 287 | ||
AK081251.1 | - | 3 | 422 | |||
AK083034.1 | - | 3 | 415 | |||
AK144551.1 | - | 3 | 548 | |||
AK201213.1 | - | 1 | 81 | |||
BC137574.1 | - | 2 | 338 | 8860593 | ||
BC137575.1 | - | 2 | 338 | 8860594 | ||
BY727964.1 | - | 3 | 427 | |||
X84239.1 | - | 2 | 295 |