Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160411.1 | NP_001153883.1 | 1 | 60 | ||
NM_026764.3 | NP_081040.1 | 2 | 265 | |||
GenBank mRNA | AF464943.1 | - | 2 | 95 | ||
AF501320.1 | - | 2 | 81 | |||
AK003418.1 | - | 1 | 61 | |||
BC030444.1 | - | 2 | 212 | 5373062 | ||
BY289676.1 | - | 2 | 265 | |||
CK335113.1 | - | 2 | 250 |