Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023456.2 | NP_075945.1 | 3, 4 | 350 | ||
GenBank mRNA | AF273768.1 | - | 2, 3 | 277 | ||
AK002982.1 | - | 3, 4 | 363 | |||
BC043012.1 | - | 3, 4 | 350 | 5683102 |