Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025156.2 | NP_001020327.1 | 3 | 323 | ||
NM_029955.3 | NP_084231.2 | 3 | 323 | |||
GenBank mRNA | AK014621.1 | - | 3 | 297 | ||
AK020306.1 | - | 3 | 283 | |||
AK052300.1 | - | 3 | 288 | |||
AK082581.1 | - | 3 | 316 | |||
AK085701.1 | - | 3 | 282 | |||
AK142302.1 | - | 3 | 341 | |||
AK159415.1 | - | 3 | 291 | |||
BC054083.1 | - | 3 | 226 | 6467830 | ||
BY136404.1 | - | 3 | 323 | |||
BY730410.1 | - | 3 | 341 |