Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027192.2 | NP_081468.1 | 5 | 983 | ||
GenBank mRNA | AK010374.1 | - | 3 | 359 | ||
BC018513.1 | - | 5 | 986 | 3597859 | ||
BC024414.1 | - | 5 | 955 | 4973810 |