Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Idi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145360.2 | NP_663335.2 | 2, 3 | 572 | ||
GenBank mRNA | AK029302.1 | - | 2, 3 | 205 | ||
AK084208.1 | - | 2, 3 | 202 | |||
AK132073.1 | - | 2, 3 | 205 | |||
AK160832.1 | - | 2, 3 | 206 | |||
BC004801.1 | - | 1, 2 | 193 | 3589498 | ||
BC110313.1 | - | 2, 3 | 210 | 5343431 | ||
CT010310.1 | - | 1, 2 | 150 |