Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C030006K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145472.2 | NP_663447.1 | 1, 2 | 194 | ||
GenBank mRNA | AK053546.1 | - | 1, 2 | 172 | ||
AK075645.1 | - | 1, 2 | 190 | |||
AK163615.1 | - | 1, 2 | 177 | |||
AK165937.1 | - | 1, 2 | 188 | |||
BC019462.1 | - | 1, 2 | 170 | 4458439 | ||
BC026997.1 | - | 1, 2 | 168 | 5362226 |