Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133786.3 | NP_598547.1 | 3 | 303 | ||
GenBank mRNA | AJ534940.2 | - | 2 | 179 | ||
AK088350.1 | - | 3 | 286 | |||
AK088846.1 | - | 3 | 269 | |||
AK133817.1 | - | 3 | 293 | |||
AK152804.1 | - | 3 | 330 | |||
BC061481.1 | - | 3 | 278 | 6512832 | ||
BC062939.1 | - | 3 | 303 | 6841276 |