Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Idi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145360.2 | NP_663335.2 | 3, 4 | 665 | ||
GenBank mRNA | AK029302.1 | - | 3, 4 | 298 | ||
AK084208.1 | - | 3, 4 | 295 | |||
AK132073.1 | - | 3, 4 | 299 | |||
AK160832.1 | - | 3, 4 | 299 | |||
BC004801.1 | - | 2, 3 | 286 | 3589498 | ||
BC110313.1 | - | 3, 4 | 303 | 5343431 | ||
CT010310.1 | - | 2, 3 | 243 |