Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145829.2 | NP_665828.1 | 6 | 1424 | ||
GenBank mRNA | AF462069.1 | - | 6 | 1380 | ||
AJ489814.1 | - | 6 | 1350 | |||
AK075765.1 | - | 5 | 1070 | |||
BC036161.1 | - | 3 | 2075 | |||
BC057990.1 | - | 6 | 1388 | 5100021 |