Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162904.1 | NP_001156376.1 | 3 | 400 | ||
NM_001162905.1 | NP_001156377.1 | 3 | 400 | |||
NM_010785.2 | NP_034915.2 | 3 | 400 | |||
NM_148922.3 | NP_683724.2 | 3 | 400 | |||
GenBank mRNA | AK004789.2 | - | 3 | 355 | ||
AK011769.1 | - | 3 | 355 | |||
BC052680.1 | - | 3 | 350 | |||
BC137608.1 | - | 3 | 328 | 8860627 | ||
BC144777.1 | - | 3 | 328 | 9053325 | ||
EU490427.1 | - | 3 | 242 | |||
EU490428.1 | - | 3 | 242 | |||
M20823.1 | - | 3 | 263 | |||
M20824.1 | - | 3 | 263 |