Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145829.2 | NP_665828.1 | 6 | 1488 | ||
GenBank mRNA | AF462069.1 | - | 6 | 1444 | ||
AJ489814.1 | - | 6 | 1414 | |||
AK075765.1 | - | 5 | 1134 | |||
BC036161.1 | - | 3 | 2139 | |||
BC057990.1 | - | 6 | 1452 | 5100021 |