- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 2 | 712 | ||
NM_001318368.1 | NP_001305297.1 | 2 | 712 | |||
XM_005266216.4 | XP_005266273.1 | 2 | 70 | |||
XM_011534864.1 | XP_011533166.1 | 2 | 691 | |||
GenBank mRNA | AK296875.1 | - | 1 | 55 | ||
AY702976.1 | - | 2 | 174 | |||
BC069634.1 | - | 2 | 197 | 7262421 | ||
BC094784.1 | - | 2 | 712 | |||
BC104492.1 | - | 2 | 200 | 40032854 | ||
BC105950.1 | - | 2 | 200 | 40032852 | ||
BC107601.1 | - | 2 | 199 | 40032849 | ||
X85753.1 | - | 2 | 200 |