Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243199.1 | NP_001230128.1 | 3 | 192 | ||
NM_001243200.1 | NP_001230129.1 | 3 | 192 | |||
NM_001243201.1 | NP_001230130.1 | 3 | 192 | |||
NM_174990.4 | NP_778155.2 | 3 | 192 | |||
NM_175048.3 | NP_778213.2 | 3 | 192 | |||
GenBank mRNA | AA543119.1 | - | 3 | 166 | ||
AA795946.1 | - | 3 | 166 | |||
AK088228.1 | - | 3 | 189 | |||
BC005577.1 | - | 3 | 182 | 3968418 | ||
BC098226.1 | - | 2 | 167 | |||
BY214550.1 | - | 3 | 193 | |||
CA576036.1 | - | 3 | 180 | |||
CB232465.1 | - | 3 | 223 |