Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290377.1 | NP_001277306.1 | 4, 5 | 740 | ||
NM_175632.3 | NP_783440.1 | 4, 5 | 758 | |||
GenBank mRNA | AF391159.1 | - | 4, 5 | 716 | ||
AF391160.1 | - | 4, 5 | 740 | |||
AF391161.1 | - | 4, 5 | 734 | |||
BC137776.1 | - | 4, 5 | 685 | 8860795 |