Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113246.2 | NP_001106717.2 | 9 | 1105 | ||
NM_029716.3 | NP_083992.1 | 2 | 306 | |||
NM_175752.3 | NP_786928.2 | 3 | 917 | |||
GenBank mRNA | AB264773.1 | - | 2590 | |||
AB264774.1 | - | 1 | 707 | |||
AF332069.1 | - | 3 | 425 | |||
AF332070.1 | - | 3 | 425 | |||
AK007182.1 | - | 2 | 306 | |||
AK032630.1 | - | 3 | 875 | |||
AK049943.1 | - | 3 | 861 | |||
AK075606.1 | - | 3 | 879 | |||
AK147686.1 | - | 3 | 861 | |||
BC010825.1 | - | 3 | 892 | 4219273 | ||
BC024796.1 | - | 3 | 868 | 5360438 | ||
BC025023.1 | - | 3 | 848 | 5364554 | ||
BC054770.1 | - | 9 | 857 | 6415204 |