Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177380.3 | NP_796354.1 | 4 | 381 | ||
GenBank mRNA | AB039380.1 | - | 4 | 381 | ||
BC125522.1 | - | 4 | 343 | 40130037 |