Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175136.2 | NP_780345.1 | 3, 4 | 535 | ||
GenBank mRNA | AK077856.1 | - | 3, 4 | 555 | ||
AK085376.1 | - | 3, 4 | 517 | |||
AK136206.1 | - | 3, 4 | 553 | |||
AK171823.1 | - | 3, 4 | 555 | |||
BC038399.1 | - | 3, 4 | 535 | 4947596 | ||
BC050268.1 | - | 2, 3 | 222 |