Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: INPPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001567.3 | NP_001558.3 | 7 | 971 | ||
XM_005273978.3 | XP_005274035.1 | 8 | 1348 | |||
XM_005273979.4 | XP_005274036.1 | 8 | 1195 | |||
XM_011544999.2 | XP_011543301.1 | 8 | 1129 | |||
XM_011545000.2 | XP_011543302.1 | 8 | 1195 | |||
XM_017017673.1 | XP_016873162.1 | 9 | 1286 | |||
GenBank mRNA | BC140853.1 | - | 7 | 971 | 9021721 | |
L36818.1 | - | 6 | 948 | |||
Y14385.1 | - | 7 | 1035 |