Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159883.1 | NP_001153355.1 | 8 | 795 | ||
NM_001159884.1 | NP_001153356.1 | 8 | 743 | |||
NM_001159885.1 | NP_001153357.1 | 7 | 778 | |||
NM_020266.2 | NP_064662.2 | 7 | 741 | |||
NM_178055.4 | NP_835156.1 | 8 | 795 | |||
GenBank mRNA | AB028858.1 | - | 7 | 608 | ||
AK076061.1 | - | 8 | 731 | |||
AK156083.1 | - | 8 | 764 | |||
AK171494.1 | - | 8 | 741 | |||
BC011090.1 | - | 8 | 697 | 3968468 | ||
BC085162.1 | - | 4 | 3090 |