Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290377.1 | NP_001277306.1 | 5 | 794 | ||
NM_175632.3 | NP_783440.1 | 5 | 812 | |||
GenBank mRNA | AF391159.1 | - | 5 | 770 | ||
AF391160.1 | - | 5 | 794 | |||
AF391161.1 | - | 5 | 788 | |||
BC137776.1 | - | 5 | 739 | 8860795 |