Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113246.2 | NP_001106717.2 | 11 | 1315 | ||
NM_001166603.1 | NP_001160075.1 | 10 | 1141 | |||
NM_001166604.1 | NP_001160076.1 | 4 | 953 | |||
NM_029716.3 | NP_083992.1 | 4 | 516 | |||
NM_175752.3 | NP_786928.2 | 5 | 1127 | |||
GenBank mRNA | AB264771.1 | - | 4 | 777 | ||
AB264772.1 | - | 10 | 838 | |||
AB264773.1 | - | 3484 | ||||
AB264774.1 | - | 3 | 917 | |||
AF332069.1 | - | 5 | 635 | |||
AF332070.1 | - | 5 | 635 | |||
AK007182.1 | - | 4 | 516 | |||
AK032630.1 | - | 5 | 1085 | |||
AK049943.1 | - | 5 | 1071 | |||
AK075606.1 | - | 5 | 1089 | |||
AK147686.1 | - | 5 | 1071 | |||
BC010825.1 | - | 5 | 1102 | 4219273 | ||
BC024796.1 | - | 5 | 1078 | 5360438 | ||
BC025023.1 | - | 5 | 1058 | 5364554 | ||
BC054770.1 | - | 11 | 1067 | 6415204 |