Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178909.4 | NP_849240.1 | 3 | 403 | ||
GenBank mRNA | AK033294.1 | - | 3 | 349 | ||
AK047691.1 | - | 3 | 378 | |||
AK088218.1 | - | 3 | 378 | |||
AK142228.1 | - | 3 | 349 | |||
BC096540.1 | - | 3 | 332 | 6403681 |